N-(2-Hydroxyethyl)-4-(1H-pyrazol-1-yl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
046573-500MG
046573-1G
046573-5G
101876-100EA
278.46
USD
101876-100
101876-102
101876-104
N-(2-Hydroxyethyl)-4-(1H-pyrazol-1-yl)benzenecarboxamide ≥95%
N-(2-Hydroxyethyl)-4-(1H-pyrazol-1-yl)benzenecarboxamide
Hazard: Irritant
Formula:
C₁₂H₁₃N₃O₂ Melting Pt: 156…158 °C |
MDL Number:
MFCD09607919 CAS Number: 1017782-46-5 |
Specification Test Results
Melting point | 156-158°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...