2-(Methylsulfonyl)-1-[4-(methylsulfonyl)phenyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

046571-500MG 046571-1G 046571-5G
101876-092EA 181.54 USD
101876-092 101876-094 101876-096
2-(Methylsulfonyl)-1-[4-(methylsulfonyl)phenyl]-1-ethanone ≥95%
2-(Methylsulfonyl)-1-[4-(methylsulfonyl)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₀H₁₂O₅S₂
Melting Pt: 207…209 °C
MDL Number: MFCD09607917
CAS Number: 952183-32-3

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR