3-(2-Fluorophenyl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluorobenzoylacetonitrile

046570-500MG 046570-1G 046570-5G
101876-086EA 278.46 USD
101876-086 101876-088 101876-090
3-(2-Fluorophenyl)-3-oxopropanenitrile ≥95%
3-(2-Fluorophenyl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₉H₆FNO
MW: 163.15 g/mol
Melting Pt: 60…62 °C
MDL Number: MFCD02260800
CAS Number: 31915-26-1

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR