1-(4-Methylbenzyl)-1H-1,2,3-triazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
046569-500MG
046569-1G
101876-082EA
556.92
USD
101876-082
101876-084
1-(4-Methylbenzyl)-1H-1,2,3-triazole-4-carbaldehyde ≥95%
1-(4-Methylbenzyl)-1H-1,2,3-triazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₁N₃O Melting Pt: 72…74 °C |
MDL Number:
MFCD09607916 CAS Number: 952183-31-2 |
Specification Test Results
Melting point | 72-74°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...