N-(2-Hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

046564-500MG 046564-1G 046564-5G
101876-060EA 278.46 USD
101876-060 101876-062 101876-064
N-(2-Hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide ≥95%
N-(2-Hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
Melting Pt: 175…177 °C
MDL Number: MFCD09607912
CAS Number: 952183-27-6

Order Now

Specification Test Results

Melting point 175-177°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR