2-Chloro-N-(2-hydroxyethyl)-4-(1H-pyrazol-1-yl)benzamide ≥95%

Supplier: Matrix Scientific

046558-500MG 046558-1G
101876-038EA 375.38 USD
101876-038 101876-040
2-Chloro-N-(2-hydroxyethyl)-4-(1H-pyrazol-1-yl)benzamide ≥95%
2-Chloro-N-(2-hydroxyethyl)-4-(1H-pyrazol-1-yl)benzamide

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃O₂
Melting Pt: 132…134 °C
MDL Number: MFCD09865002
CAS Number: 1172238-65-1

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR