2-Chloro-N-(2-hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
046556-500MG
046556-1G
101876-028EA
375.38
USD
101876-028
101876-030
2-Chloro-N-(2-hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide ≥95%
2-Chloro-N-(2-hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide
Hazard: Irritant
Formula:
C₁₃H₁₇ClN₂O₂ Melting Pt: 104…106 °C |
MDL Number:
MFCD09607906 CAS Number: 952183-24-3 |
Specification Test Results
Melting point | 104-106°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...