(2-Phenyl-1H-imidazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

046554-500MG 046554-1G
101876-020EA 241.54 USD
101876-020 101876-022
(2-Phenyl-1H-imidazol-4-yl)methanol ≥95%
(2-Phenyl-1H-imidazol-4-yl)methanol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
Melting Pt: 144…146 °C
MDL Number: MFCD06202903
CAS Number: 43002-54-6

Order Now

Specification Test Results

Melting point 144-146°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR