3-(4-Ethylphenyl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

046552-500MG 046552-1G 046552-5G
101876-010EA 278.46 USD
101876-010 101876-012 101876-014
3-(4-Ethylphenyl)-3-oxopropanenitrile ≥95%
3-(4-Ethylphenyl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO
Melting Pt: 105…107 °C
MDL Number: MFCD02260779
CAS Number: 96220-15-4

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR