3-Amino-3-(4-ethylphenyl)acrylonitrile 95
Supplier: Matrix Scientific
046550-500MG
046550-1G
046550-5G
101876-000EA
181.54
USD
101876-000
101876-002
101876-004
3-Amino-3-(4-ethylphenyl)acrylonitrile 95
3-Amino-3-(4-ethylphenyl)acrylonitrile
Hazard: Irritant
Formula:
C₁₁H₁₂N₂ MW: 172.23 g/mol Melting Pt: 88…90 °C |
MDL Number:
MFCD09607902 CAS Number: 952183-22-1 |
Specification Test Results
Melting point | 88-90°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...