5-Chloro-2-pyridinethiol ≥95%

Supplier: Matrix Scientific

046549-1G 046549-5G
101875-996EA 192.31 USD
101875-996 101875-998
5-Chloro-2-pyridinethiol ≥95%
5-Chloro-2-pyridinethiol

Hazard: Irritant

Formula: C₅H₄ClNS
Melting Pt: 195…198 °C
MDL Number: MFCD00661468
CAS Number: 40771-41-3

Order Now

Specification Test Results

Melting point 195-198°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR