(2-Methyl-1H-imidazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Hydroxymethyl)-2-methyl-1H-imidazole

046548-500MG 046548-1G
101875-992EA 1109.23 USD
101875-992 101875-994
(2-Methyl-1H-imidazol-4-yl)methanol ≥95%
(2-Methyl-1H-imidazol-4-yl)methanol

Hazard: Irritant

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD06202901
CAS Number: 45533-87-7

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR