Ethyl-5-methyl-1,1-dioxo-1,6-dihydro-1-λ~6~2,6-thiadiazine-3-carboxylate ≥95%
Supplier: Matrix Scientific
046540-500MG
046540-1G
046540-5G
101875-974EA
375.38
USD
101875-974
101875-976
101875-978
Ethyl-5-methyl-1,1-dioxo-1,6-dihydro-1-λ~6~2,6-thiadiazine-3-carboxylate ≥95%
Ethyl-5-methyl-1,1-dioxo-1,6-dihydro-1-lambda~6~2,6-thiadiazine-3-carboxylate
Hazard: Irritant
Formula:
C₇H₁₀N₂O₄S Melting Pt: 99…101 °C |
MDL Number:
MFCD08443997 CAS Number: 5863-20-7 |
Specification Test Results
Melting point | 99-101°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...