2-Hydrazino-6-nitrobenzothiazole ≥95%
Supplier: Matrix Scientific
046539-500MG
046539-1G
101875-970EA
556.92
USD
101875-970
101875-972
2-Hydrazino-6-nitrobenzothiazole ≥95%
2-Hydrazino-6-nitrobenzothiazole
Hazard: Irritant
Formula:
C₇H₆N₄O₂S Melting Pt: 273-274°C |
MDL Number:
MFCD00022864 CAS Number: 30710-21-5 |
Specification Test Results
Melting point | 273-274°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...