Ethyl-2-imino-3-methyl-2,3-dihydro-1,3-benzothiazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,3-dihydro-2-imino-3-methyl-1,3-benzothiazole-6-carboxylate

046538-500MG 046538-1G 046538-5G
101875-964EA 278.46 USD
101875-964 101875-966 101875-968
Ethyl-2-imino-3-methyl-2,3-dihydro-1,3-benzothiazole-6-carboxylate ≥95%
Ethyl-2-imino-3-methyl-2,3-dihydro-1,3-benzothiazole-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂S
MW: 236.29 g/mol
Melting Pt: 273…274 °C
MDL Number: MFCD08443996
CAS Number: 924868-95-1

Order Now

Specification Test Results

Melting point 273-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR