Ethyl-2-chloro-6-benzothiazolecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloro-6-benzothiazolecarboxylate

046537-500MG 046537-1G 046537-5G
101875-958EA 156.92 USD
101875-958 101875-960 101875-962
Ethyl-2-chloro-6-benzothiazolecarboxylate ≥97%
Ethyl-2-chloro-6-benzothiazolecarboxylate

Hazard: Irritant

Formula: C₁₀H₈ClNO₂S
MW: 241.6900 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD08443995
CAS Number: 78485-37-7

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR