N-(2-Chloroethyl)-N'-(4-pyridinyl)urea ≥95%

Supplier: Matrix Scientific

046535-500MG 046535-1G 046535-5G
101875-946EA 375.38 USD
101875-946 101875-948 101875-950
N-(2-Chloroethyl)-N'-(4-pyridinyl)urea ≥95%
N-(2-Chloroethyl)-N'-(4-pyridinyl)urea

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MW: 199.64 g/mol
Melting Pt: 102…106 °C
MDL Number: MFCD01676543
CAS Number: 62491-96-7

Order Now

Specification Test Results

Melting point 102-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR