1-Boc-4-(2-chloroethyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Chloroethyl)piperazine,N1-BOC protected, tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate

046534-1G 046534-5G
101875-942EA 153.85 USD
101875-942 101875-944
1-Boc-4-(2-chloroethyl)piperazine ≥95%
1-Boc-4-(2-chloroethyl)piperazine

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O₂
MW: 248.751 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD08443993
CAS Number: 208167-83-3

Order Now

Specification Test Results

Melting point 67-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR