1-Boc-4-(2-hydroxyethyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 4-(2-Hydroxyethyl)piperazine, N1-BOC protected

046530-500MG 046530-1G 046530-5G
101875-916EA 181.54 USD
101875-916 101875-918 101875-920
1-Boc-4-(2-hydroxyethyl)piperazine ≥95%
1-Boc-4-(2-hydroxyethyl)piperazine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₃
Boiling Pt: 140°C/1 Torr
Melting Pt: 41…43 °C
MDL Number: MFCD00728947
CAS Number: 77279-24-4

Order Now

Specification Test Results

Melting point 41-43°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR