2',4'-Dihydroxy-3'-[(dimethylamino)methyl]acetophenone ≥95%

Supplier: Matrix Scientific

046528-500MG 046528-1G 046528-5G
101875-910EA 181.54 USD
101875-910 101875-912 101875-914
2',4'-Dihydroxy-3'-[(dimethylamino)methyl]acetophenone ≥95%
2',4'-Dihydroxy-3'-[(dimethylamino)methyl]acetophenone

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
Melting Pt: 106-108°C
MDL Number: MFCD08443990
CAS Number: 924868-94-0

Order Now

Specification Test Results

Melting point 106-108°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR