Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
046527-500MG
046527-1G
046527-5G
101875-904EA
210.77
USD
101875-904
101875-906
101875-908
Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate ≥95%
Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₃ MW: 220.23 g/mol Melting Pt: 219…221 °C |
MDL Number:
MFCD08443989 CAS Number: 627901-54-6 |
Specification Test Results
Melting point | 219-221°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...