Ethyl-4-(4,4-dimethylpiperidin-1-yl)benzoate ≥95%
Supplier: Matrix Scientific
046526-500MG
046526-1G
046526-5G
101875-898EA
375.38
USD
101875-898
101875-900
101875-902
Ethyl-4-(4,4-dimethylpiperidin-1-yl)benzoate ≥95%
Ethyl-4-(4,4-dimethylpiperidin-1-yl)benzoate
Hazard: Irritant
Formula:
C₁₆H₂₃NO₂ Melting Pt: 88…90 °C |
CAS Number:
406233-25-8 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...