Ethyl-4-(2-hydroxy-5-methyl-3-nitrophenyl)-2,4-dioxobutanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,4-dioxo-4-(2-hydroxy-5-methyl-3-nitrophenyl)butanoate

046525-500MG 046525-1G 046525-5G
101875-892EA 278.46 USD
101875-892 101875-894 101875-896
Ethyl-4-(2-hydroxy-5-methyl-3-nitrophenyl)-2,4-dioxobutanoate ≥95%
Ethyl-4-(2-hydroxy-5-methyl-3-nitrophenyl)-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₁₃H₁₃NO₇
MW: 295.25 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD08443987
CAS Number: 859843-60-0

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR