4-[(Dimethylamino)methyl]-2,6-diisopropylbenzenol ≥95%
Supplier: Matrix Scientific
046524-500MG
046524-1G
046524-5G
101875-886EA
278.46
USD
101875-886
101875-888
101875-890
4-[(Dimethylamino)methyl]-2,6-diisopropylbenzenol ≥95%
4-[(Dimethylamino)methyl]-2,6-diisopropylbenzenol
Hazard: Irritant
Formula:
C₁₅H₂₅NO Melting Pt: 84…86 °C |
MDL Number:
MFCD00514869 CAS Number: 4918-95-0 |
Specification Test Results
Melting point | 84-86°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...