tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
3-[(Dimethylamino)methylene]piperidin-4-one, N1-BOC protected
046523-500MG
046523-1G
046523-5G
101875-880EA
100
USD
101875-880
101875-882
101875-884
tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₂₂N₂O₃ MW: 254.33 g/mol Melting Pt: 85…87 °C |
MDL Number:
MFCD08443986 CAS Number: 157327-41-8 |
Specification Test Results
Melting point | 85-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...