Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(4,4-dimethyl-2,6-dioxopiperidin-1-yl)benzoate

046521-500MG 046521-1G 046521-5G
101875-868EA 278.46 USD
101875-868 101875-870 101875-872
Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate ≥95%
Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₉NO₄
MW: 289.33 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD01764136
CAS Number: 279692-23-8

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR