Methyl-4-({[(2-chloroethyl)amino]carbonyl}amino)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

046518-500MG 046518-1G 046518-5G
101875-850EA 278.46 USD
101875-850 101875-852 101875-854
Methyl-4-({[(2-chloroethyl)amino]carbonyl}amino)benzenecarboxylate ≥95%
Methyl-4-({[(2-chloroethyl)amino]carbonyl}amino)benzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
MW: 256.69 g/mol
Melting Pt: 165…166 °C
MDL Number: MFCD03149047
CAS Number: 380324-25-4

Order Now

Specification Test Results

Melting point 165-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR