1-(4-Methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3,5-Dimethyl-1-(4-methoxyphenyl)-1H-pyrazole-4-carboxylic acid
046515-500MG
046515-1G
046515-5G
101875-838EA
375.38
USD
101875-838
101875-840
101875-842
1-(4-Methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(4-Methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₃ MW: 246.27 g/mol Melting Pt: 180…183 °C |
MDL Number:
MFCD08310579 CAS Number: 1001779-87-8 |
Specification Test Results
Melting point | 180-183°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...