2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylbenzenol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylphenol
046513-500MG
046513-1G
046513-5G
101875-828EA
278.46
USD
101875-828
101875-830
101875-832
2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylbenzenol ≥95%
2-(tert-Butyl)-6-[(dimethylamino)methyl]-4-methylbenzenol
Hazard: Irritant
Formula:
C₁₄H₂₃NO MW: 221.35 g/mol Melting Pt: 46…49 °C |
MDL Number:
MFCD08443981 CAS Number: 4142-59-0 |
Specification Test Results
Melting point | 46-49°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...