Ethyl-2-methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3,4-dihydro-2-methyl-7-nitro-3-oxo-2H-1,4-benzoxazine-2-carboxylate

046512-500MG 046512-1G 046512-5G
101875-822EA 278.46 USD
101875-822 101875-824 101875-826
Ethyl-2-methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%
Ethyl-2-methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₆
MW: 280.24 g/mol
Melting Pt: 193…195 °C
MDL Number: MFCD08443980
CAS Number: 154365-36-3

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR