Ethyl-2-methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%
Supplier: Matrix Scientific
046511-500MG
046511-1G
046511-5G
101875-816EA
278.46
USD
101875-816
101875-818
101875-820
Ethyl-2-methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%
Ethyl-2-methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₆ MW: 280.24 g/mol Melting Pt: 173…174 °C |
MDL Number:
MFCD08443979 CAS Number: 154365-37-4 |
Specification Test Results
Melting point | 173-174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...