2-[5-Methyl-2-(2-thienyl)-1,3-thiazol-4-yl]acetic acid ≥95%

Supplier: Matrix Scientific

046501-500MG 046501-1G 046501-5G
101875-762EA 747.69 USD
101875-762 101875-764 101875-766
2-[5-Methyl-2-(2-thienyl)-1,3-thiazol-4-yl]acetic acid ≥95%
2-[5-Methyl-2-(2-thienyl)-1,3-thiazol-4-yl]acetic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂S₂
MW: 239.32 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD08443972
CAS Number: 924868-89-3

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR