2-{5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%
Supplier: Matrix Scientific
046500-500MG
046500-1G
046500-5G
101875-756EA
375.38
USD
101875-756
101875-758
101875-760
2-{5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%
2-{5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Hazard: Irritant
Formula:
C₁₃H₁₀F₃NO₂S MW: 301.29 g/mol Melting Pt: 154…156 °C |
MDL Number:
MFCD08443971 CAS Number: 924868-88-2 |
Specification Test Results
Melting point | 154-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...