2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%

Supplier: Matrix Scientific

046498-500MG 046498-1G 046498-5G
101875-744EA 375.38 USD
101875-744 101875-746 101875-748
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₂S
MW: 301.29 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD08443969
CAS Number: 924868-87-1

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR