4-(3-Bromobenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046497-500MG 046497-1G
101875-740EA 375.38 USD
101875-740 101875-742
4-(3-Bromobenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(3-Bromobenzoyl)-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₈BrNO₃
Melting Pt: 220…222 °C
MDL Number: MFCD08443968
CAS Number: 924868-86-0

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR