3-[4-(1H-Pyrazol-1-yl)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

046494-500MG 046494-1G
101875-728EA 278.46 USD
101875-728 101875-730
3-[4-(1H-Pyrazol-1-yl)phenyl]acrylic acid ≥95%
3-[4-(1H-Pyrazol-1-yl)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
Melting Pt: 233…235 °C
MDL Number: MFCD06205764
CAS Number: 1006441-28-6

Order Now

Specification Test Results

Melting point 233-235°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR