3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

046489-500MG 046489-1G 046489-5G
101875-700EA 278.46 USD
101875-700 101875-702 101875-704
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde ≥95%
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₈Cl₂O
Melting Pt: 95…98 °C
MDL Number: MFCD03002047
CAS Number: 50670-78-5

Order Now

Specification Test Results

Melting point 95-98°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR