3-Methyl-4-(2-thienyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

046488-500MG 046488-1G 046488-5G
101875-694EA 181.54 USD
101875-694 101875-696 101875-698
3-Methyl-4-(2-thienyl)-1H-pyrazole ≥95%
3-Methyl-4-(2-thienyl)-1H-pyrazole

Hazard: Irritant

Formula: C₈H₈N₂S
MW: 164.23 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD08443963
CAS Number: 1006491-93-5

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR