3-[5-(2-Chlorophenyl)-2-furyl]acrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-[5-(2-Chlorophenyl)fur-2-yl]acrylic acid

046487-500MG 046487-1G 046487-5G
101875-688EA 375.38 USD
101875-688 101875-690 101875-692
3-[5-(2-Chlorophenyl)-2-furyl]acrylic acid ≥95%
3-[5-(2-Chlorophenyl)-2-furyl]acrylic acid

Hazard: Irritant

Formula: C₁₃H₉ClO₃
MW: 248.67 g/mol
Melting Pt: 192…194 °C
MDL Number: MFCD00695964
CAS Number: 58110-39-7

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR