5-Bromo-6-(dimethylamino)-2-methyl-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
046484-500MG
046484-1G
101875-674EA
807.69
USD
101875-674
101875-676
5-Bromo-6-(dimethylamino)-2-methyl-4-pyrimidinol ≥95%
5-Bromo-6-(dimethylamino)-2-methyl-4-pyrimidinol
Hazard: Irritant
Formula:
C₇H₁₀BrN₃O Melting Pt: 178…180 °C |
MDL Number:
MFCD12025867 CAS Number: 1135283-29-2 |
Specification Test Results
Melting point | 178-180°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...