Methyl-2-nitro-5-piperidinobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

046483-500MG 046483-1G
101875-670EA 278.46 USD
101875-670 101875-672
Methyl-2-nitro-5-piperidinobenzenecarboxylate ≥95%
Methyl-2-nitro-5-piperidinobenzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₄
Melting Pt: 117…119 °C
MDL Number: MFCD12025866
CAS Number: 773071-23-1

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR