5-Bromo-2-methyl-6-(methylamino)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046482-500MG 046482-1G
101875-666EA 747.69 USD
101875-666 101875-668
5-Bromo-2-methyl-6-(methylamino)-4-pyrimidinol ≥95%
5-Bromo-2-methyl-6-(methylamino)-4-pyrimidinol

Hazard: Irritant

Formula: C₆H₈BrN₃O
Melting Pt: 225…227 °C
MDL Number: MFCD12025865
CAS Number: 1135283-28-1

Order Now

Specification Test Results

Melting point 225-227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR