3-Bromo-1-methyl-1H-indole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046480-500MG 046480-1G
101875-660EA 515.38 USD
101875-660 101875-662
3-Bromo-1-methyl-1H-indole-2-carboxylic acid ≥95%
3-Bromo-1-methyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
Melting Pt: 179 °C
MDL Number: MFCD11099656
CAS Number: 499983-77-6

Order Now

Specification Test Results

Melting point 206-208°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR