1-Methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

046476-500MG 046476-1G
101875-644EA 800 USD
101875-644 101875-646
1-Methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylic acid ≥95%
1-Methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₆F₃NO₂
Melting Pt: 210…212 °C
MDL Number: MFCD12025862
CAS Number: 130530-01-7

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR