Ethyl-3-amino-5-bromo-1-benzofuran-2-carboxylate ≥95%
Supplier: Matrix Scientific
046475-500MG
046475-1G
101875-640EA
509.23
USD
101875-640
101875-642
Ethyl-3-amino-5-bromo-1-benzofuran-2-carboxylate ≥95%
Ethyl-3-amino-5-bromo-1-benzofuran-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀BrNO₃ Melting Pt: 155…157 °C |
MDL Number:
MFCD00454292 CAS Number: 330555-71-0 |
Specification Test Results
Melting point | 155-157°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...