3-(1-Benzofuran-2-yl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

046470-500MG 046470-1G
101875-616EA 376.92 USD
101875-616 101875-618
3-(1-Benzofuran-2-yl)-3-oxopropanenitrile ≥95%
3-(1-Benzofuran-2-yl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₁H₇NO₂
Melting Pt: 149…151 °C
MDL Number: MFCD00047286
CAS Number: 5149-69-9

Order Now

Specification Test Results

Melting point 149-151°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR