3-(5-Bromo-1-benzofuran-2-yl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

046469-500MG 046469-1G
101875-612EA 423.08 USD
101875-612 101875-614
3-(5-Bromo-1-benzofuran-2-yl)-3-oxopropanenitrile ≥95%
3-(5-Bromo-1-benzofuran-2-yl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₁H₆BrNO₂
Melting Pt: 163…165 °C
MDL Number: MFCD12025858
CAS Number: 1135283-25-8

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR