3-Oxo-3-(4-piperidinophenyl)propanenitrile ≥95%
Supplier: Matrix Scientific
046468-500MG
046468-1G
101875-608EA
375.38
USD
101875-608
101875-610
3-Oxo-3-(4-piperidinophenyl)propanenitrile ≥95%
3-Oxo-3-(4-piperidinophenyl)propanenitrile
Hazard: Irritant
Formula:
C₁₄H₁₆N₂O MW: 228.3 g/mol Melting Pt: 152…154 °C |
MDL Number:
MFCD12025857 CAS Number: 1135283-23-6 |
Specification Test Results
Melting point | 152-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...