3-Oxo-3-[4-(1-pyrrolidinyl)phenyl]propanenitrile ≥95%
Supplier: Matrix Scientific
046467-500MG
046467-1G
101875-604EA
423.08
USD
101875-604
101875-606
3-Oxo-3-[4-(1-pyrrolidinyl)phenyl]propanenitrile ≥95%
3-Oxo-3-[4-(1-pyrrolidinyl)phenyl]propanenitrile
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O MW: 214.27 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD07786223 CAS Number: 887595-01-9 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...