3-(4-Morpholinophenyl)-3-oxopropanenitrile ≥95%
Supplier: Matrix Scientific
046464-500MG
046464-1G
101875-600EA
375.38
USD
101875-600
101875-602
3-(4-Morpholinophenyl)-3-oxopropanenitrile ≥95%
3-(4-Morpholinophenyl)-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₂ MW: 230.27 g/mol Melting Pt: 118…120 °C |
MDL Number:
MFCD07787044 CAS Number: 887591-40-4 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...